Learn More
Electron Microscopy Sciences Forceps Warmer
SDP
Supplier:  Electron Microscopy Sciences 6320120
Encompass_Preferred
This forceps warmer is indestructible: An all metal, space saving unit. Three heated wells prevent wax build-up and allow for easy tissue orientation at the embedding centers.
Catalog No. 50-259-88
Explore available promotions