Learn More
Yamato Scientific America, Inc. Yamato 90L Forced Convection Oven, Programmable, 220V
Supplier: Yamato Scientific America, Inc. DKN412C
Programmable 90L forced convection constant temperature oven with extended functions and safety features. Bestseller based on excellent performance and affordability. Superior temperature accuracy. Comes with observation window. Programmable PID controller for easy program settings (30-step, 3-pattern program controller with repeat functions). Fixed setting, Quick Auto stop, Auto stop, Auto start, Program operating modes with easy control capabilities. Overheating prevention and calibration off-set are possible with auxiliary functions. Easy to use and maintain. Equipped with built in exhaust ports. Operating temp ambient +10~260°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.