Learn More
Scientific Industries FORCE GAU TORBAL FB1K 200LBF
Supplier: Scientific Industries FB1K
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
200lbf x 0.05lbf (1kN x 0.2N) The FB1k is an affordable, fully configurable force gauge designed for a wide range of tension and compression applications. The force gauge is intuitive and easy-to-use with logically arranged user menus and ready for out-of-the-box use, no complicated configuration or setup. USB, RS232, and microSD allow for easy PC connectivity and data management. The FBk digital force gauge is rugged and ergonomic with an auto rotating backlit LCD display which automatically adjusts to the orientation of the unit. The FBk includes all essential force measuring modes such as Peak, Multi Peak, and Standard (Real Time). Every function is optimized with statistical analysis, onscreen graphs, and a histogram. Easy-to-use Pass / Fail threshold settings makes this force gauge ideal for quality control applications. The FBk series is equipped with four powerful and easily removable batteries which keep the unit charged up to 32 hours.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.