Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Force Biphenyl LC Columns, Particle Size: 1.8 μm
Ideal for bioanalytical testing applications such as drug and metabolite analyses
$819.00 - $955.00
Specifications
For Use With | Bioanalytical Testing Applications |
---|---|
Ligand Type | Biphenyl |
Max. Pressure | 830 bar (12,000 psi) |
Particle Size | 1.8 μm |
pH | 2 to 8 |
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
06-611-542
|
Restek™
9629232 |
30 mm | 2.1 mm |
Each for $819.00
|
|
||||
06-611-543
|
Restek™
9629252 |
50 mm | 2.1 mm |
Each for $881.00
|
|
||||
06-611-544
|
Restek™
962925E |
50 mm | 3 mm |
Each for $881.00
|
|
||||
06-611-540
|
Restek™
9629212 |
100 mm | 2.1 mm |
Each for $955.00
|
|
||||
06-611-541
|
Restek™
962921E |
100 mm | 3 mm |
Each for $925.00
|
|
||||
Description
- Increased retention for dipolar, unsaturated, or conjugated solutes
- Enhanced selectivity when used with methanolic mobile phase
- Ideal for increasing sensitivity and selectivity in LC-MS analyses
- Heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries
- Limits ionization suppression and allows simple, MS-friendly mobile phases
- Restek’s most popular LC stationary phase
Specifications
Bioanalytical Testing Applications | |
830 bar (12,000 psi) | |
2 to 8 | |
300 m2/g | |
L11 | |
Fully Porous Silica | |
Yes |
Biphenyl | |
1.8 μm | |
100 Å | |
80°C | |
Phenyl | |
15% | |
HPLC/UHPLC Column |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title