Learn More
Labconco™ FlaskScrubber™ Laboratory Glassware Washer Accessory, Pipet Inserts
For use with FlaskScrubber and FlaskScrubber Vantage Series Laboratory Glassware Washers.
$746.00 - $782.00
Specifications
Material | Stainless Steel (304) |
---|---|
Product Type | Pipette Insert |
For Use With (Application) | Washing and drying pipets |
For Use With (Equipment) | FlaskScrubber and FlaskScrubber Vantage Series Glassware Washers |
Description
- Wash and dry pipets inside and out
- Allows direct injection cleaning of pipets
- Self-sealing silicone check valves allow washing of partial or full loads
- May be placed in lower Spindle Rack in Labconco FlaskScrubber or FlaskScrubber Vantage Series
- May fit up to 4 Inserts at once in FlaskScrubber or FlaskScrubber Vantage Series washers
Warranty and Services
1 year
Specifications
Stainless Steel (304) | |
Pipette Insert | |
Washing and drying pipets | |
FlaskScrubber and FlaskScrubber Vantage Series Glassware Washers |