Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Food Coloring in Dropper Bottles for Science Kits
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Consumable used with multiple projects and in science kits.
Description
- Bottles contains 30mL (1 oz.) of food coloring
Specifications
Red Food Coloring | |
Multiple projects or science kits | |
Each | |
30 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction