Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-Trp(Boc)-SerPsi(Me,Me)Pro-OH | 908601-15-0, 10GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1198810GM
Encompass
WuXi TIDES Fmoc-Trp(Boc)-SerPsi(Me,Me)Pro-OH, 908601-15-0 | 3-(Na-(((9H-fluoren-9-yl)methoxy)carbonyl)-1-(tert-butoxycarbonyl)tryptophyl)-2,2-dimethyloxazolidine-4-carboxylic acid | CC(C)(C)OC(=O)N1C=C(CC(NC(=O)OCC2C3=CC=CC=C3C4=C2C=CC=C4)C(=O)N5C(C(=O)O)COC5(C)C)C6=C1C=CC=C6 | M.F. C37H39N3O8, M.W. 653.73
Catalog No. 50-234-2034