Learn More
Agilent Technologies FMOC REAGENT, 2.5 MG/ML-10X1ML
Supplier: Agilent Technologies 50613337
FMOC reagent, 2.5 mg/mL in acetonitrile, 9-fluorenylmethylchloroformate, 10 x 1 mL. Dangerous goods, 10X1ML
This item is not returnable.
View return policy