Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-L-Tyr(2,6-dichloro-Bzl)-OH | 112402-12-7, 100GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA12043100GM
Encompass
WuXi TIDES Fmoc-L-Tyr(2,6-dichloro-Bzl)-OH, 112402-12-7 | (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((2,6-dichlorobenzyl)oxy)phenyl)propanoic acid | ClC1=CC=CC(Cl)=C1COC=2C=CC(=CC2)C[C@@H](C(O)=O)NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O | M.F. C31H25Cl2NO5, M.W. 562.44
Catalog No. 50-234-3315