Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-L-Arg(Pbf)-Opfp | 200132-16-7, 10GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1186610GM
Encompass
WuXi TIDES Fmoc-L-Arg(Pbf)-Opfp, 200132-16-7 | perfluorophenyl N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-Nw-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)-L-argininate | CC1=C2C(CC(C)(O2)C)=C(C(S(=O)(NC(NCCC[C@H](NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O)C(OC6=C(F)C(F)=C(F)C(F)=C6F)=O)=N)=O)=C1C)C | M.F. C40H39F5N4O7S, M.W. 814.83
Catalog No. 50-234-2571