Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-alpha-methyl-D-4-Fluorophe | 1217777-84-8, 50GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1148450GM
Encompass
WuXi TIDES Fmoc-alpha-methyl-D-4-Fluorophe, 1217777-84-8 | (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-fluorophenyl)-2-methylpropanoic acid | C[C@@](C(O)=O)(NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)CC4=CC=C(F)C=C4 | M.F. C25H22FNO4, M.W. 419.45
Catalog No. 50-234-4446