Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-5-Hydroxy-DL-tryptophan | 351857-99-3, 10GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1144010GM
Encompass
WuXi TIDES Fmoc-5-Hydroxy-DL-tryptophan, 351857-99-3 | 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-hydroxy-1H-indol-3-yl)propanoic acid | OC=1C=CC2=C(C1)C(=CN2)CC(C(O)=O)NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O | M.F. C26H22N2O5, M.W. 442.47
Catalog No. 50-234-4864