Learn More
Biocare Medical, LLC Fluorescence Antibody Diluent - 100 ml
Supplier: Biocare Medical, LLC FAD901L
Fluorescence antibody diluent is a modified PBS formulation with protein carrier and preservative. This ready-to-use modified PBS is formulated to deliver superior performance and signal preservation. The diluent is protease free and stabilizes fluorescent dyes for up to one month in a prediluted format.