Learn More
Biotium Fluorescein-DHPE
Supplier: Biotium 60024
Fluorescein-DHPE (N-(Fluorescein-5-thiocarbamoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt) is a useful surface pH indicator and has also been used for measuring lateral diffusion in membranes via fluorescence photobleaching recovery method. Properties: Ex/Em(MeOH) = 496/ 519nm; Orange solid soluble in chloroform; Store at -20°C and protect from light; C64H100N3O13PS; MW: 1182.53; [87706-98-7]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.