Learn More
Cole-Parmer Fluke 720RTD RTD Probe for 721 and 719Pro
Supplier:  Cole-Parmer 1610797
Encompass_Preferred
Expand the capabilities of your calibrator with the temperature probe and adapter. Protect your calibrator using the liquid trap.
Catalog No. NC2223155