Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Supplier: Thermo Scientific™ R118821
Description
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
- For sterility testing according to USP (United States Pharmacopeia)
Specifications
Industrial Reference: AOAC, BAM, COMPF, USP. | |
20mm x 113mm Tube | |
Microbiology Media |
Fluid Thioglycollate Medium | |
United States | |
20/Pk. |