Learn More
Fluotics Fluent Evo MCA comp P50 non-sterile
Supplier: Fluotics MCA50.NS
Tecan Fluent Evo MCA compatible P50, non-sterile, for MCA head, working volume: 50 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30038606
This item is not returnable.
View return policy