Learn More
Cytiva (Formerly Pall Lab) Metricel Polypropylene Membrane Disc Filters - 25 mm (100/pkg)
Supplier: Cytiva M5PU025
Pure Polypropylene Membrane for Filtration of Aggressive Solvents-Pure polypropylene gives high chemical stability-Offers high temperature resistance and low extractable levels This product is not approved for use in medical, clinical, surgical or other patient applications.