Learn More
Cytiva (Formerly Pall Lab) Open-face Filter Holders - 47 mm, open-face aluminum (1/pkg)
Supplier: Cytiva 1220
The open-face filter holders are lightweight and corrosion-resistant.Available in three sizes: 25, 37, and 47 mmIncludes capsThe 25 mm holder is made of Delrin plastic and is designed for clean room sampling, breathing-zone air samples collecting, bacterial monitoring, particulate analysis, etc.The 37 and 47 mm holders are constructed of aluminum. They can be used for trapping airborne particles, fluorescent tracers, and other contaminants. They include a plastic cap that protects samples after filtration. This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.