Learn More
Cytiva (Formerly Pall Lab) 47 mm In-line Filter Holder, Aluminum - 47 mm, in-line aluminum (1/pkg)
Supplier: Cytiva 1235
The 47 mm filter holders are designed for in-line liquid or gas filtration. They open and close easily without disturbing the filter disc.Designed for in-line liquid or gas filtrationEasy open and closeAvailable in a variety of materialsThey are available in polycarbonate and aluminum.For venting and air monitoring applications, the sturdy plastic construction of the lightweight polycarbonate 47 mm in-line filter holder is ideal.For stack sampling applications, the 47 mm in-line filter holder in anodized aluminum is a durable and cost-effective choice.For excellent chemical resistance and continuous processing of fluids at high pressure, the 47 mm in-line filter holder in stainless steel is recommended. This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.