Learn More
Chemglass Life Sciences 500mL Flask, Reaction, 24/40 Outer Joint, 2mm Glass Stopcock, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF052008
Encompass_Preferred
500mL Flask, Reaction, 24/40 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids. Lower portion of flask fits standard heating mantles. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. AF052008