Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter, Correlated Reading, PTFE Housing and Fittings, 150-mm Scale; 374 mL/min Air
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0326762
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.