Learn More
Dwyer Instruments Inc Flow Meter, 2"""" Scale, 10-100 CC/Min Air, SS Valve
Supplier: Dwyer Instruments Inc RMA150SSV
Accurate within ±4% of full scale reading. Scales are brushed aluminum, coated with epoxy and the graduations are on both sides of the indicating tube.