Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ FlexTech™ DuPont™ Aramid Fiber Gloves with Foam Nitrile Coating
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
Description
- Lightweight stretch Aramid Fiber provides flexibility and cut protection
- Foam nitrile palm enhances grip and resistance to snags and abrasions
- Uncoated back circulates air for breathability to keep hands cool and comfortable
- Non-slip surface increases efficiency and worker confidence
Specifications
ANSI Abrasion Rating | ANSI 4 |
Certifications/Compliance | ASTM 1790-97, ASTM F2992-15, ASTM D3389-05, ASTM F1358-08 |
Coating Material | Nitrile Foam |
Color | Yellow |
Cut Resistance Rating | ANSI A2 |
Hand | Specific |
Material | Aramid Fiber |
Product Type | Cut-resistant Gloves |
Size | X-Large |
EN388 Standard | EN 388:3532 |
Show More |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction