Learn More
Glas Col LLC Evaporator
Supplier: Glas Col LLC 109A YH112
Evaporator FlexiVap workstation with heating for small insert. 24 positions with flowmeter. Heating to 100 degrees C maximum. 120V, 50 psi.