Learn More
Cole-Parmer Oakton Flexible Benchtop Electrode Holder
Supplier: Cole-Parmer 3561750
Holds up to four standard size electrodes (14-mm dia max) and one ATC probe. Weighted 6'' (15.2 cm) diameter base lets you pivot holder 360 for added convenience. Flexible arm lets you move electrode side to side or up and down while keeping electrode at a constant vertical angle.