Learn More
Perkin Elmer US LLC Flat Sample Disk - Tin, Qty. 1000
Supplier: Perkin Elmer US LLC N2410244
The Flat Sample Disk is made of a minimum of 99.0% pure tin. It is 1.1 inches in diameter, and used to analyzing 13 mm or 25 mm filters. These glass filters are commonly used in air pollution and oceanographic studies for quantitating air or ocean particulates.