Learn More
Electron Microscopy Sciences Flat Emery Stick 3/0 Grit
Supplier: Electron Microscopy Sciences 6211903
Emery paper which is glued to a wooden handle. 11" long x 3/4" wide (279x19.1 mm). Very useful for filing excess resin of sample block tips.