Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ FlaskScrubber™ Vantage™ Series Laboratory Glassware Washer Accessory, Small Spindles w/Clips



For use with FlaskScrubber and FlaskScrubber Vantage Series Laboratory Glassware Washers
Manufacturer: Labconco™ 4596000P/EMD
Description
- Included with every FlaskScrubber and FlaskScrubber Vantage series glassware washer
- Provide direct injection cleaning of small narrowneck glassware on spindle sacks
- Spindles may be used in any spindle location on any upper or lower spindle rack
- May be used in place of lower spindle rack in any FlaskScrubber or FlaskScrubber Vantage Series washer
Specifications
I.D.: 12 in. | |
Small Spindle with Clip | |
Package of 10 small spindles with 10 height adjustment clips, tool to remove large spindles, 36 nylon plugs and 18 height adjustment clips for large spindles | |
10.9 cm |
Stainless Steel (304) | |
FlaskScrubber and FlaskScrubber Vantage Series Laboratory Glassware Washer | |
4.3 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title