Learn More
Chemglass Life Sciences 200mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask
Supplier: Chemglass Life Sciences CG151259
200mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
This item is not returnable.
View return policy