Learn More
Wilmad Labglass FLASK FAT EXTRACTION 35MM OD
Supplier: Wilmad Labglass LG6912100
Flask, Fat Extraction, AOAC. Capacity: 110mL; Body OD: 35mm; Stopper Size: no. 1. For the extraction of fat from milk products. Reference AOAC 989.05. Supplied w/out stopper.