Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI FlashPure Chromatography Cartride Sample Packs
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries for your purification needs.
$421.00 - $440.00
Specifications
Type | Chromatography Cartridge |
---|---|
Includes | Sample Bundle of Cartridges |
Column Type | Stationary Phase |
Catalog Number | Mfr. No. | Bed Weight (Metric) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight (Metric) | Material | Price | Quantity & Availability | ||||
01-670-195
|
BUCHI
34004989 |
12 g, 20 g, 80 g | C18 |
Pack for $440.00
|
|
||||
01-670-196
|
BUCHI
34004990 |
4 g, 12 g, 20 g, 25 g, 40 g, 80 g | Silica, C18 |
Pack for $421.00
|
|
||||
Description
FlashPure EcoFlex cartridges are designed with optimized backpressure, price and performance.
- Normal Phase Bundle for basic applications and polar compounds
- Reverse Phase C18 Bundle for nonpolar compounds and ideal for peptides
- Scale-up Bundle for larger loading separations
Specifications
Chromatography Cartridge | |
Sample Bundle of Cartridges | |
Stationary Phase |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title