Learn More
Cole-Parmer FF-200D-I Economical Flame Photometer; 230 VAC, 50/60 Hz
Supplier: Cole-Parmer 8305505
Use this economical flame photometer for routine determinations of sodium and potassium in industrial and educational applications. This easy-to-maintain unit gives you fast and accurate results. For safety, an automatic gas shutoff mechanism activates if the flame becomes accidentally extinguished. Unit comes with one each of Na+, K+, Li+, Ca+, and Ba+ filters, two meters each of gas and air tubing, and necessary fittings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.