Learn More
Fisherbrand™ Syringe Filters: Nylon Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
$319.50 - $4060.00
Catalog Number | Diameter (Metric) | Pore Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Diameter (Metric) | Pore Size | Quantity | Price | Quantity & Availability | ||||
09-719-007
|
33 mm | 0.45 μm | 50/Pk. |
Pack of 50 for $319.50
|
|
||||
09-719-008
|
33 mm | 0.45 μm | 150/Pk. |
Pack of 150 for $661.00
|
|
||||
09-719-009
|
33 mm | 0.45 μm | 1500/Pk. |
Pack of 1500 for $4,060.00
|
|
||||
09-719-006
|
33 mm | 0.2 μm | 150/Pk. |
Pack of 150 for $616.00
|
|
||||
09-720-000
|
13 mm | 0.22 μm | 100/Pk. |
Pack of 100 for $351.00
|
|
||||
09-720-001
|
13 mm | 0.45 μm | 100/Pk. |
Pack of 100 for $351.00
|
|
||||
Description
Low hold-up volume helps you to recover virtually all of your sample after filtering
Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.
Nylon Membrane
- Excellent chemical resistance and low protein binding
- Ideal for aqueous/organic/HPLC applications
- Hydrophilic
Specifications
Nylon | |
Non-sterile | |
Syringe Filter |