Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ SURE-TEK™ 2 Processing and Embedding Cassette Taped Sleeves
Used in Leica Cassette Labelers
$337.00
Specifications
For Use With (Application) | Leica Cassette Labelers |
---|---|
Type | Processing and Embedding Cassette Taped Sleeve |
Catalog Number | Color | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Includes | Price | Quantity & Availability | |||||
22-048-102
|
Blue | For table or wall mounting |
Case of 1000 for $337.00
|
|
22-048-101
|
White | Attached lid |
Case of 1000 for $337.00
|
|
22-048-103
|
Green | Attached lid |
Case of 1000 for $337.00
|
|
22-048-106
|
Peach | Attached lid |
Case of 1000 for $337.00
|
|
22-048-107
|
Yellow | Attached lid |
Case of 1000 for $337.00
|
|
22-048-109
|
Aqua | Attached lid |
Case of 1000 for $337.00
|
|
22-048-110
|
Lilac | Attached lid |
Case of 1000 for $337.00
|
|
22-048-104
|
Gray | Attached lid | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
22-048-105
|
Pink | Attached lid | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-048-108
|
Tan | Attached lid | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
Description
- Comes with attached lids
- Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac color
Specifications
Leica Cassette Labelers | |
Processing and Embedding Cassette Taped Sleeve |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title