Learn More
Fisherbrand™ Superfrost™ Excell™ Microscope Slides
New, innovative adhesive technology for superior adhesion and wettability
Description
New, innovative adhesive technology for superior adhesion and wettability
- Developed specifically for immunohistochemistry laboratory applications
- Adhesion surface for heat-induced epitope retrieval
- For use in HIER methods requiring high pH antigen retrieval solutions, including EDTA
- Uniform hydrophilic surface works well with automated staining equipment
- Constructed of glass to work well with both epon and methyl methacrylate sections
- Hydrophilic surface allows sections to spread well during initial attachment; section adherence is maintained throughout difficult staining procedures
- Excellent tissue adherence of all tissue types, yet easily release identified areas for Laser Capture Microdissection
- Enhanced attachment of anchorage dependent cell lines, including many fastidious lines, e.g., primary neuronal cells
Specifications
Yes | |
Glass | |
Clipped |
Microscope Slide | |
Adhesion |