Learn More
Fisherbrand™ Poxygrid™ Test Tube Racks
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
 View more versions of this product
Catalog No. 14-793-14
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
Autoclavable
Tubes (10 to 13 mm)
72
3
8.1 x 4.4 x 2.5 in. (20.5 x 11.3 x 6.4cm)
8.1 in.
4.4 in.
2.5 in.
13 mm
Blue
Epoxy Coated Steel
Rectangle
Test Tube Rack
20.5 cm
11.3 cm
6.4 cm
13 mm