Learn More
Fisherbrand™ Hanhart 112.0101-00 Crown Stopper Mechanical Stopwatch
Crown Stopper Mechanical Stopwatch
Supplier: Fisherbrand™ 112.0101
Description
7 Jewel mechanical stopwatch with crown stopper, pin lever with shock-protected balance wheel, 30 min. Register and 60 sec. sweep hand, 0.2 sec. recorder, unbreakable spring and an alloy case.
- 1/5 second recorder
- 30 min. register
- 60 second sweep
- Shock protected
- Pinlever
- 7 jewel
- Alloy brass case
Specifications
7 Jewel Mechanical Stopwatch | |
30 min. | |
Metal | |
Crown stopper, pin lever with shock-protected balance wheel, 30 min. Register and 60 sec. sweep hand, 0.2 sec. recorder, 7 jewels with an unbreakable spring and an Alloy case |