Learn More
Fisher Healthcare™ Corrugated Slide Mailers/Holder
Patented window design for easy viewing of the patient's name
Description
- Double well on the top and bottom of the mailer allows room for the slide to dry when mailer is closed
- Unique tab closure system eliminates the need for tape
- Holds one or two 3 x 1 in. slides
Specifications
Double Slide Mailer | |
Corrugated Carton | |
1000/Cs. |