Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ ColorFrost™ Microscope Slides with Clipped Corners
Durable, opaque printed surface inert to common laboratory chemicals and reagents
$170.00 - $191.00
Specifications
Frosted | Colorfrost |
---|---|
Thickness (Metric) | 1 mm |
Length (Metric) | 25 mm |
Width (Metric) | 75 mm |
Catalog Number | Color | Corner Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Corner Style | Price | Quantity & Availability | |||||
22-037-160
|
Blue | Clipped |
Pack of 144 for $191.00
Case of 10 PK for $1,637.00
|
|
|||||
22-037-161
|
Green | Clipped |
Pack of 144 for $170.00
Case of 10 PK for $1,540.00
|
|
|||||
22-037-162
|
Pink | 45° Clipped |
Pack of 144 for $191.00
Case of 10 PK for $1,642.00
|
|
|||||
22-037-163
|
Yellow | Clipped |
Pack of 144 for $191.00
Case of 10 PK for $1,641.00
|
|
|||||
Description
- 45° clipped corners; color-coded frosted end
- Raised printed writing surface eliminates need for paper labels and tedious relabeling of slides
- Autoclavable
Specifications
Colorfrost | |
1 mm | |
25 mm | |
75 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title