Learn More
Fisherbrand™ Sequencing Gel Caster
Bubble-free sequencing gels in less than three minutes
Catalog No. FB-SEQ-CST
Add to cart
Includes: Adjustable casting platform, width adjustment knobs, and 12 binder clamps
Description
Description
  • Simple, innovative, and quick method for casting
  • No taping or special assembly is required
  • System is adjustable—accommodates glass plates from 20 to 42cm wide and 48cm long
  • Horizontal plate sliding, capillary action drawing gel between plates, and surface tension keep gel from leaking
Specifications
Specifications
12.2 cm
71 cm
Sequencing Gel Caster
21 cm
7.8 to 16.5 in.
20 to 42 cm
25.7 in.