Learn More
Traceable™ One-Shot™ pH Buffer Standards pH Certified Reference Material (CRM), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
$35.30
Specifications
For Use With (Equipment) | pH Meter and Electrode |
---|---|
Quantity | 6 x 100 mL |
Solution Type | Buffer |
Traceability to NIST | Certified and Traceable™ |
Catalog Number | Chemical Name or Material | pH | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Chemical Name or Material | pH | Packaging | Price | Quantity & Availability | ||||
15-078-287
|
Buffer Standards One-Shot pH 10.012 (CRM) | 10.012 | 100 mL One-Shot (6 Pk.) |
Pack of 6 for $35.30
|
|
||||
15-078-285
|
Buffer Standards One-Shot pH 4.005 (CRM) | 4.005 | 100 mL One-Shot (6 Pk.) |
Pack of 6 for $35.30
|
|
||||
15-078-286
|
Buffer Standards One-Shot pH 7.000 (CRM) | 7.00 | 100 mL One-Shot (6 Pk.) |
Pack of 6 for $35.30
|
|
||||
15-078-288
|
One-Shot pH Buffer Standards (CRM) 6-Pack Assortment | 4.005, 7.000, 10.012 | 100 mL One-Shot (2 of each) |
Pack of 6 for $35.30
|
|
||||
Description
Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied with individual temperature chart, traceability information and Traceable™ Certificate
Specifications
pH Meter and Electrode | |
6 x 100 mL | |
Buffer | |
Certified and Traceable™ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.