Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Isotemp™ Flammable-Materials Storage Refrigerator/Freezers
For storage of volatile materials
Description
Safely store ethyl ether, hexane, naphtha, benzol, epoxies, solvents, fuels, pharmaceuticals, clinical reagents, and biologicals.
- Manual defrost
- CFC-free refrigerant
- Foamed polyurethane insulation ensures temperature uniformity
- White enamel steel exteriors
- ABS white plastic interiors
- White epoxy-coated steel shelves
- Doors open from left
- Orange warning label reads: “Flammable Material Storage Refrigerator”
- Sparkproof interiors separate contents from compressor and motor
- Top-mounted hydraulic thermostat and door stop
- Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
- Compressors have enclosed overloads, relays, and thermostats to prevent sparks from escaping during opening and closing
- Static condensers and evaporators are free of electric fans
Two-door 10 cu. ft. (283L) Unit
- Refrigerator: 2° to 10°C, 8.0 cu. ft. (226.5L)
- Freezer: -18° to -5°C, 2.0 cu. ft. (56.6L) Five white epoxy-coated steel adjustable shelves, four fixed shelves, one full-width slide-out bottom drawer in refrigerator
- One fixed shelf, one door compartment in freezer
- Two adjustable and two fixed pads allow easy installation and leveling
Order Info
For 115V 60Hz. Supplied with NEMA 5-15P plug; requires NEMA 5-15R wall receptacle.
Service Plan
Service agreement available.
Certifications
UL listed, CSA approved
Compliance
Complies with OSHA 29CFR1910.106 and NFPA Standard Numbers 45, 56C, and 99.
Warranty and Services
13 months, parts and labor
Specifications
Flammable Material Storage Freezer/Refrigerator | |
8.0 cu. ft. | |
-12°C to -20°C | |
Hinged, Open Left to Right, Reversible | |
3 Adjustable, 1 Fixed, 1 Drawer | |
Bottom-mounted | |
R134a, 100% CFC Free | |
Enameled Steel | |
ABS Plastic | |
2 Adjustable Feet In Front, 2 Fixed Pads In Rear | |
Mechanical Dial Thermostat/Outside on Rear of Unit | |
13 Months Parts and Labor | |
65 in. | |
24.5 in. | |
23.5 in. | |
1.9 A | |
115 V | |
60 Hz | |
170 lb. |
10 cu. ft. | |
226.5 L | |
1°C to 12°C | |
2 (2 compartments) | |
4 | |
Automatic (Refrigerator), Manual (Freezer) | |
Steel | |
Polyurethane Foam | |
NFPA 45 and 70, UL | |
White | |
10FCEEFSA | |
24.5 x 23.5 x 65 in. (62 x 60 x 165 cm) | |
165 cm | |
62 cm | |
60 cm | |
NEMA 5-15 P | |
115 V 60 Hz | |
230 w | |
77 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction