Learn More
Fisherbrand™ Isotemp™ CO2 Backup Systems for Ultra-Low Temperature Freezers, CO2 backup system, field installed
For use with Isotemp Upright and Chest Ultra-Low Temperature Freezers
Description
For use with Isotemp Upright and Chest Ultra-Low Temperature Freezers
Maintains temperature down to -70°C
Specifications
CO2 Backup System | |
-70°C | |
Upright | |
Isotemp™ Ultra low temperature freezer |