Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Science Superhero Tote Bag
Spacious, multipurpose two-toned canvas tote bag featuring the STEMcredibles superheroes.
Description
- L x W x H: 18 x 7 x 14 in.
- 22 in. double stitched handles
- Snap closure
- Front procket
- White
- Kid friendlty/CPSIA compliant
Specifications
Tote bag | |
White, Blue | |
18 x 7 x 14 in. (45.7 x 17.8 x 35.5cm) | |
45.7 cm | |
17.8 cm | |
35.5 cm |
22 in. double stitched handles | |
Snap cloure and front pocket | |
18 in. | |
7 in. | |
14 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction