Learn More
Fisher Science Education™ Advanced Digital Stereomicroscopes
SCI_ED
Extra-rugged stereomicroscopes have a built-in high-resolution digital camera.
Supplier:  Fisher Science Education™ DC5420TH
 View more versions of this product
Catalog No. S71013
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes: Motic™ Images software, which allows the user to view, capture, label, manipulate and measure images; vinyl dustcover; rubber eyeshields
Description
Description
College/University Level
  • Versatile stereomicroscope permits user to change from a widefield image at 10X magnification to close-up detail simply by rotating the objective turret
  • Microscope is fully functional, independent of the camera
  • S71013 features a 3.0 MP live image
Warranty and Services
Lifetime limited warranty on microscope portion, excluding bulbs, camera and cords; Camera warranty: one year
Specifications
Specifications
10X to 40X
Digital Stereo Microscope
Binocular, Inclined/Rotating
Incidental/Transmitted
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories