Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Sure-Vue™ Signature Rubella Latex Test
Does not require a dilution step for 10 IU sensitivity
Specifications
CPT Code | 86762, 86406 |
---|---|
Clia Complexity | Non-waived |
Detectable Analytes | Anti-rubella virus IgG, IgM |
Type | Rubella Latex Test Kit |
Includes | Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards and package insert |
Includes: Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards, and package insert
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
23-038-060
|
100 Tests |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View alternate product |
N/A | ||||||
23-038-062
|
500 Tests |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View alternate product |
N/A | ||||||
Description
- CLIA Complexity: Non-Waived
- CPT Codes: 86762/86406
- Sensitivity: 100%
- Specificity: 98.6%
- Rapid latex agglutination test for the qualitative and semi-quantitative detection of both IgG and IgM anti-rubella virus antibodies in serum
- Results in five minutes
- For in vitro diagnostic use
Specifications
86762, 86406 | |
Anti-rubella virus IgG, IgM | |
Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards and package insert |
Non-waived | |
Rubella Latex Test Kit | |
5 min. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title