Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Sure-Vue™ Color Mono Test Kit
Ideal for standardization
Specifications
Clia Complexity | Non-waived |
---|---|
Detectable Analytes | Infectious Mononucleosis Heterophil Antibodies |
Type | Color Mononucleosis Latex Test Kit |
Includes | Reagent, disposable slides and prediluted positive and negative control |
Includes: Reagent, disposable slides, prediluted positive and negative controls, and package insert
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
23-038017
|
50 Tests |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View Alternate Product |
N/A | ||||||
Description
- CLIA Complexity: Non-Waived
- CPT Code: 86308
- Color-enhanced slide test designed for detection of infectious mononucleosis heterophile antibodies in serum or plasma
- Test results in two minutes
- Sensitivity: 97.9%
- Specificity: 95.8%
- For in vitro diagnostic use
Specifications
Non-waived | |
Infectious Mononucleosis Heterophil Antibodies | |
Color Mononucleosis Latex Test Kit | |
Reagent, disposable slides and prediluted positive and negative control |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title