Learn More
Fisher Healthcare™ Sure-Vue™ Adeno Test Kit
Simple two-step procedure
 View more versions of this product
Catalog No. SA067022
Due to product restrictions, please to purchase or view availability for this product.
Includes: 20 Test devices, tubes containing extraction buffer, disposable sample transfer pipets, package insert
Description
Description
  • CLIA Complexity: Moderate
  • CPT Code: 87301
  • Results in 15 minutes
  • Rapid, membrane-based immunogold assay for the qualitative detection of adenovirus and its antigens
  • Aids in the diagnosis of conjunctivitis, respiratory illnesses and gastroenteritis caused by adenovirus
  • Can be used with eye swabs, nasopharyngeal secretions, fecal samples, and cell culture supernatant
  • Positive and negative controls sold separately
Specifications
Specifications
87301
Qualitative detection of Adenovirus and its Antigens in eye swabs, nasal and pharyngeal secretions and fecal material
2 Vials
Adenovirus Testing Control Set
20 Test devices, Tubes containing extraction Buffer, disposable sample transfer pipets, package insert