Description
- Bovine serum albumin (BSA) commonly used as an additive or nutrient in cell and microbial culture
- Free of DNase, RNase, and protease
- BSA is used to stabilize enzymes during DNA digestion
- Can also be used to help determine quantity of unknown proteins in protein assays
- BSA in water with protein concentration of 20 mg/mL
Specifications
20mg/mL | |
BSA Solution | |
Bovine | |
Solution |
Protein Stabilizer and Blocking Agent | |
5mL | |
Amber Glass |